Skip to Content
Merck
All Photos(1)

Documents

DLW354775

Corning® PureCoat Plate 24w Carboxyl 5pac 5cas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107

shipped in

wet ice

storage temp.

room temp

Legal Information

Corning is a registered trademark of Corning, Inc.
PureCoat is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service