Skip to Content
Merck
All Photos(1)

Documents

CRMUPPLU

Pour Point Standard

IP15, ASTM D97, ISO 3016, BS 2000, Pt 15

Synonym(s):

IP15, ASTM D97, Pt 15 Pour Point Standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

Agency

according to ASTM® D97
according to BS 2000
according to ISO 3016

manufacturer/tradename

Paragon Scientific Ltd

bp

175-270 °C

mp

-58 °C

application(s)

petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service