Skip to Content
Merck
All Photos(1)

Key Documents

CRM8033

Supelco

ICP-MS Alternate Internal Standard 2

certified reference material, 100 μg/mL each component in 10 wt. % nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

bottle of 125 mL

concentration

100 μg/mL each component in 10 wt. % nitric acid

format

multi-component solution

storage temp.

room temp

General description

May be used to monitor and correct for changes that occur from differences between standards and samples. Since environmental samples often contain significant amounts of lithium, isotopically enriched 95% 6Li can be analyzed as an internal standard, avoiding the signal from the 7Li peak.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analyte

Description

    Bi 100 μg/mL
    Ge 100 μg/mL
    In 100 μg/mL
    6Li 100 μg/mL
    Lu 100 μg/mL
    Rh 100 μg/mL
    Sc 100 μg/mL
    Tb 100 μg/mL
    See All (8)

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service