Skip to Content
Merck
All Photos(1)

Key Documents

CCW4721

Millipore

Coliforms Chromogenic Agar Plates

Solid, selective, and differential culture medium for the detection and enumeration of total coliform and E. coli in water samples by the membrane-filtration technique acc. to ISO, suitable for microbiology

Synonym(s):

CCA

Sign Into View Organizational & Contract Pricing

Select a Size

30 EACH
€96.80

€96.80


Available to ship onApril 07, 2025Details


Request more information

Select a Size

Change View
30 EACH
€96.80

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€96.80


Available to ship onApril 07, 2025Details


Request more information

Agency

ISO 9308-1:2014

Quality Level

form

solid

shelf life

5 mo. (Expiry date on the label)

feature

ready-to-use

composition

agar, 13.00 g/L
5-bromo-4-chloro-3-indoxyl-β-D-glucuronic acid, 0.10 g/L
6-chloro-3-indoxyl-β-D-galactopyranoside, 0.10 g/L
di-sodium hydrogen phosphate, 2.70 g/L
enzymatic digest of casein, 1.00 g/L
IPTG, 0.10 g/L
sodium chloride, 5.00 g/L
Sodium dihydrogen phosphate dihydrate, 2.20 g/L
sodium pyruvate, 1.00 g/L
sorbitol, 1.00 g/L
Tergitol 7, 0.15 g/L
tryptophan, 1.00 g/L
yeast extract, 2.00 g/L

packaging

box of 30 plates

storage condition

(Store dry and tightly closed)

parameter

9 mL filling volume

plate diam.

55 mm

color

pale yellow

pH

6.8±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

2-25°C

suitability

Escherichia coli
coliforms

General description

Coliforms Chromogenic Agar Plates are ready-to-use plates to detect and enumerate total coliform and E. coli in water samples. The combined action of peptone, pyruvate, and sorbitol allows rapid colony growth in this phosphate-buffered medium, which also permits simple recovery of sublethal thermally injured coliforms. Sodium chloride provides the correct osmotic environment necessary for growth. The selectivity is attained partially by the Tergitol 7, which inhibits the growth of Gram-positive and some Gram-negative bacteria without affecting the coliform bacteria. The culture medium is formulated without antibiotics for water samples with low bacterial background flora. The colonial differentiation is due to the chromogenic mixture, composed of two enzyme substrates: 6-chloro-3-indoxyl-ß-D-galacto-pyranoside (Salmon-GAL) and 5-bromo-4-chloro-3-indoxyl-ß-D-glucuronide (X-Glucuronide). ß-D-galactosidase in coliforms cleaves the first substrate, giving a salmon-red color to the coliform colonies. The ß-D-glucuronidase enzyme, characteristic of E. coli, cleaves the second chromogenic substrate, giving blue color to their colonies.E. coli has two enzymes that cleave both chromogenic substances giving dark blue to violet colonies. Total coliforms are the sum of E. coli colonies and the salmon-red colonies. The IPTG enhances the metabolism of chromogens. Other Gram-negative bacteria produce colorless colonies. However, some Gram-negative bacteria possessing glucuronidase activity (not galactosidase) produce light blue to turquoise colonies.E. coli colonies are confirmed by including a small amount of tryptophane in the medium and coating the blue-violet colonies with a drop of Kovacs Reagent. The color change of the reagent to cherry-red in a few seconds confirms the production of indole and thereby, the presence of E. coli. When the Chromogenic Agar for Coliform is used with the membrane filter method, the color and growth of the colonies can be modified by the characteristics of the membrane filter. It is advisable to perform validation of the membrane filter type used.

Application

Coliforms Chromogenic Agar Plates are suitable for the detection and enumeration of total coliform and E. coli in water samples by the membrane-filtration technique.

Components

1 box containing: 6 plastic bags with 5 plates of 55 mm/bag

Other Notes

If the membrane filtration method is used, it must be considered that the nature and characteristics of the filter membrane used also influences the size and color of the colonies grown on this culture medium.

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service