Skip to Content
Merck
All Photos(1)

Documents

BP409

Methysergide maleate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Methysergide maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O2 · C4H4O4
CAS Number:
Molecular Weight:
469.53
MDL number:
UNSPSC Code:
41116107

grade

pharmaceutical primary standard

API family

methysergide

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.[H][C@@]12Cc3cn(C)c4cccc(C1=C[C@H](CN2C)C(=O)N[C@@H](CC)CO)c34

InChI

1S/C21H27N3O2.C4H4O4/c1-4-15(12-25)22-21(26)14-8-17-16-6-5-7-18-20(16)13(10-23(18)2)9-19(17)24(3)11-14;5-3(6)1-2-4(7)8/h5-8,10,14-15,19,25H,4,9,11-12H2,1-3H3,(H,22,26);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,15+,19-;/m1./s1

InChI key

LWYXFDXUMVEZKS-ZVFOLQIPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methysergide maleate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service