Skip to Content
Merck
All Photos(1)

Key Documents

BCR545

Welding dust loaded on filter [Cr(VI), Cr]

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€298.00

€298.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
€298.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€298.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Packaging

glass fibre filter, loaded with dust containing about 100 μg Cr (VI

Analysis Note

For more information please see:
BCR545

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service