Skip to Content
Merck
All Photos(1)

Key Documents

BCR487

Pig liver (vitamins)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

shipped in

wet ice

storage temp.

−20°C

Analysis Note

For more information please see:
BCR487

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Vitamins: 5-Methyltetrahydrofolic acid (5-MTHF), Folate (total), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), Vitamin B12, Vitamin B6 (total pyridoxine)

Matrix Group: Meat
Sales restrictions may apply

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niladri Sekhar Chatterjee et al.
Journal of AOAC International, 100(3), 579-585 (2017-03-18)
Despite the potential of LC with tandem MS (MS/MS) in improving sensitivity and selectivity, analytical methods are scarce for the determination of protein-bound and phosphorylated forms of B vitamins in food. This prompted us to develop a method for LC-MS/MS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service