Skip to Content
Merck
All Photos(4)

Documents

B8630

Sigma-Aldrich

Bromothymol Blue sodium salt

powder

Synonym(s):

3′,3′′-dibromothymolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27Br2NaO5S
CAS Number:
Molecular Weight:
646.36
Beilstein:
6887926
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

technique(s)

titration: suitable

visual transition interval

6.0-7.6, yellow to blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CC(C)c1cc(c(C)c(Br)c1O)C(=C2\C=C(C(C)C)C(=O)C(Br)=C2C)\c3ccccc3S([O-])(=O)=O

InChI

1S/C27H28Br2O5S.Na/c1-13(2)18-11-20(15(5)24(28)26(18)30)23(17-9-7-8-10-22(17)35(32,33)34)21-12-19(14(3)4)27(31)25(29)16(21)6;/h7-14,30H,1-6H3,(H,32,33,34);/q;+1/p-1/b23-21+;

InChI key

NMKFVGALBGZKGW-FKWCIMQXSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Bromothymol Blue sodium salt has been used as a pH indicator in the yeast growth media (YPD, yeast extract peptone dextrose).

Biochem/physiol Actions

Bromothymol Blue is a pH indicator which changes color from yellow (pH 6.0) to blue (7.6).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increasing galactose consumption by Saccharomyces cerevisiae through metabolic engineering of the GAL gene regulatory network.
Ostergaard, et al.
Nature Biotechnology, 18, 1283-1286 (2000)
The roles of galactitol, galactose-1-phosphate, and phosphoglucomutase in galactose-induced toxicity in Saccharomyces cerevisiae.
de Jongh WA, et al.
Biotechnology and Bioengineering, 101, 317-326 (2008)
Handbook of Acid-Base Indicators null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service