Skip to Content
Merck
All Photos(1)

Key Documents

85549

Sigma-Aldrich

Span® 85

Synonym(s):

Sorbitan trioleate, Span® 85

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H108O8
CAS Number:
Molecular Weight:
957.49
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

refractive index

n20/D 1.476 (lit.)

viscosity

200-300 mPa.s(20 °C)

HLB value

1.8±1.1

density

0.94 g/mL at 20 °C
0.956 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H](CO)[C@H]1OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C60H108O8/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-56(62)65-52-54(61)59-60(68-58(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)55(53-66-59)67-57(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54-55,59-61H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27+/t54-,55+,59-,60-/m1/s1

InChI key

ZBNRGEMZNWHCGA-WECPCUOASA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Span is a registered trademark of Croda International PLC

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

437.0 °F

Flash Point(C)

225 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gülay Büyükköroğlu et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 109, 174-183 (2016-10-30)
Cervical cancer is one of the most life threatening types of cancer among women and is generally resistant to chemotherapy. The objective of this study was to prepare a vaginal suppository containing a chemotherapeutic agent and a genetic material that
P M Blackett et al.
Pharmaceutical research, 12(11), 1689-1693 (1995-11-01)
The purpose of the work is to study the adsorption of Oleic acid and Span 85 (materials frequently used in aerosols as surfactants) onto partially amorphous and essentially crystalline salbutamol sulphate, attempting to understand the behaviour of metered dose inhalers
Hiroaki Todo et al.
International journal of pharmaceutics, 271(1-2), 41-52 (2004-05-08)
The effect of pulmonary absorption enhancers on the stability of active ingredients is an important factor for successful inhalation therapy as well as the effect on pharmacological activity and safety. We examined the effect of pulmonary absorption enhancers on the
A N Hassan et al.
International journal of food microbiology, 87(1-2), 145-152 (2003-08-21)
The influence of surfactant hydrophobicity on detachment of Escherichia coli O157:H7 from lettuce was determined. Lettuce pieces inoculated with the pathogen were rinsed with Tween and Span surfactants of different hydrophobicity. Of the Tweens, only Tween 85, the Tween with
Maik J Geerken et al.
Journal of colloid and interface science, 312(2), 460-469 (2007-05-08)
The formation of emulsions with micro-engineered silicon based arrays of micro-orifices is a relatively new technique. Until now, only the preparation of oil-in-water emulsions was studied due to the hydrophilic nature of silicon. This work evaluates the emulsification of water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service