81542
Polystyrene A-HMPA
extent of labeling: 0.7-1.0 mmol/g loading
Synonym(s):
2-[4-(Hydroxymethyl)phenoxyacetamido]ethyl-polystyrene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
extent of labeling
0.7-1.0 mmol/g loading
matrix
crosslinked with 1% DVB
particle size
500-560 μm
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service