Skip to Content
Merck
All Photos(1)

Key Documents

75310

Millipore

OGY Agar

suitable for microbiology

Synonym(s):

Oxytetracycline Glucose Yeast-extract Agar Basis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
D(+)-glucose, 10 g/L
yeast extract, 5 g/L

final pH

6.5±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care

microbiology

suitability

selective for Aspergillus spp.
selective for Candida spp.
selective for Penicillium spp.
selective for Pichia spp.
selective for Saccharomyces spp.
selective for Zygosaccharomyces spp.
selective for molds (General Media)
selective for yeasts (General Media)

Application

Selective agar according to Mossel for the identification and enumeration of yeasts and other fungi in foods, clinical specimens etc.

Preparation Note

Supplements:
2 vial/l Oxytetra Selective Supplement (Cat. No. 51239)
may be:1 ml/l gentamicin solution (Cat. No. 48755)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxytetracycline-glucose-yeast extract agar for selective enumeration of moulds and yeasts in foods and clinical material.
D A Mossel et al.
The Journal of applied bacteriology, 33(3), 454-457 (1970-09-01)

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Yeast is one of the most important microorganisms known and utilised by mankind. Ancient Middle Eastern civilisations used the organism to bake bread and to produce mead, beer and wine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service