Skip to Content
Merck
All Photos(1)

Key Documents

69427

Supelco

1-Fluoronaphthalene

reference material

Synonym(s):

Duloxetine impurity G (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F
CAS Number:
Molecular Weight:
146.16
Beilstein:
1906413
MDL number:
UNSPSC Code:
41116107

grade

reference material

Assay

≥97.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.593 (lit.)

bp

215 °C (lit.)

mp

−13 °C (lit.)

density

1.1322 g/mL at 20 °C (lit.)

SMILES string

Fc1cccc2ccccc12

InChI

1S/C10H7F/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

CWLKTJOTWITYSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service