Skip to Content
Merck
All Photos(3)

Key Documents

62567

Supelco

Lithium ionophore VI

for ion-selective electrodes, Selectophore, function tested, ≥99.0% (HPLC)

Synonym(s):

6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane, 6,6-Dibenzyl-14-crown-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€709.00

€709.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€709.00

About This Item

Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51
Beilstein:
5991129
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

€709.00


Check Cart for Availability

Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (HPLC)

form

solid

quality

function tested

SMILES string

C1COCCOCC(COCCOC1)(Cc2ccccc2)Cc3ccccc3

InChI

1S/C24H32O4/c1-3-8-22(9-4-1)18-24(19-23-10-5-2-6-11-23)20-27-16-14-25-12-7-13-26-15-17-28-21-24/h1-6,8-11H,7,12-21H2

InChI key

KDMUFHBMXZLDLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Neutral carrier for Li+-ions in PVC ion-selective electrodes with excellent selectivities over potassium and sodium ions

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mona N Sweilam et al.
Biosensors & bioelectronics, 150, 111897-111897 (2019-12-02)
Flexible wearable chemical sensors are emerging tools which target diagnosis and monitoring of medical conditions. One of the potential applications of wearable chemical sensors is therapeutic drug monitoring for drugs that have a narrow therapeutic range such as lithium. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service