Skip to Content
Merck
All Photos(1)

Key Documents

59968

Millipore

Modified lauryl sulfate tryptose broth acc. GB

suitable for microbiological culture, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607

Agency

GB 4789.40-2016

Quality Level

form

powder

composition

Dipotassium hydrogen phosphate, 2.75 g
Lactose, 5.0 g
Potassium dihydrogen phosphate, 2.75 g
Sodium chloride, 34.0 g
Sodium lauryl sulfate, 0.1 g
Trypticase, 20.0 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.8± 0.2 (25 °C)

suitability

suitable for microbiology

application(s)

food and beverages
microbiology

Application

Recommended by GB 4789.40-2016 for selective enrichment of Cronobacter (formerly E. sakazakii) from milk and milk products.

Preparation Note

Suspend 64.6g/L, autoclave at 121 ℃ for 15 minutes, when cooling to 45-50 ℃, add 1 vial of Vancomycin Supplement (Cat. No. 75423).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service