Skip to Content
Merck
All Photos(1)

Key Documents

50992

Supelco

N-Methyl-N-trimethylsilyltrifluoroacetamide activated I

for GC derivatization, LiChropur, activated with ethanethiol and ammonium iodide

Synonym(s):

MSTFA activated I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.22
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.380

storage temp.

2-8°C

Application

  • Derivatizing agent for the trimethylsilylation of -OH and -NH groups for GC and GC-MS[1]
  • Suitable for the silylation of testosterone.

Features and Benefits

  • Activated with ethanethiol and ammonium iodide.
  • This mixture is equivalent to a 1000:2 mixture of MSTFA and iodotrimethylsilane.        

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F

Flash Point(C)

26 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W. Schanzer et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 687, 93-93 (1996)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service