Skip to Content
Merck
All Photos(1)

Key Documents

48716

Millipore

Gassner Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Water Blue Metachrome-yellow Lactose Agar according to Gassner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L
lactose, 43 g/L
metachrome yellow, 1.25 g/L
peptone, 14 g/L
sodium chloride, 5 g/L
water blue, 0.62 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages

microbiology

suitability

selective and differential for Escherichia coli
selective and differential for coliforms
selective and differential for enterobacteriaceae

Application

Selective agar for the detection and isolation of pathogenic Enterobacteriaceae in food and other material according to Gassner.

Preparation Note

Dissolve 76.87 g in 1 litre distilled water. Autoclave at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gassner, G.
Centralbl. F. Bakt. I. Orig., 80, 219-219 (1918)

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service