Skip to Content
Merck
All Photos(1)

Documents

46436

Supelco

6α-Methylprednisolone

VETRANAL®, analytical standard

Synonym(s):

11β,17α,21-Trihydroxy-6α-methyl-1,4-pregnadiene-3,20-dione, 6α-Methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, Medrol, Medrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O5
CAS Number:
Molecular Weight:
374.47
Beilstein:
2340300
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

functional group

ketone

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1

InChI key

VHRSUDSXCMQTMA-PJHHCJLFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juho Keski-Nisula et al.
The Annals of thoracic surgery, 95(6), 2126-2132 (2013-04-23)
Corticosteroids are widely used in pediatric open-heart surgery to reduce systemic inflammatory response and to mediate possible cardioprotective effects. However, the optimal dosing of corticosteroids is unknown and their administration varies considerably between different institutions. Forty neonates undergoing open-heart surgery
Christiaan Fokke et al.
Brain : a journal of neurology, 137(Pt 1), 33-43 (2013-10-29)
Guillain-Barré syndrome is an acute polyradiculoneuropathy with a variable clinical presentation. Accurate diagnostic criteria are essential for patient care and research, including clinical trials and vaccine safety studies. Several diagnostic criteria for Guillain-Barré syndrome have been proposed, including the recent
Ornulf Paulsen et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 32(29), 3221-3228 (2014-07-09)
Corticosteroids are frequently used in cancer pain management despite limited evidence. This study compares the analgesic efficacy of corticosteroid therapy with placebo. Adult patients with cancer receiving opioids with average pain intensity ≥ 4 (numeric rating scale [NRS], 0 to
Richard Whitlock et al.
American heart journal, 167(5), 660-665 (2014-04-29)
Steroids may improve outcomes in high-risk patients undergoing cardiac surgery with the use of cardiopulmonary bypass (CBP). There is a need\ for a large randomized controlled trial to clarify the effect of steroids in such patients. We plan to randomize
Emilio Portaccio et al.
Journal of neurology, neurosurgery, and psychiatry, 85(8), 845-850 (2014-01-10)
To assess relapses, disability progression and the role of disease modifying drugs (DMDs) in the year after delivery in women with multiple sclerosis (MS). We prospectively followed-up pregnancies occurring between 2002 and 2008 in women with MS, recruited from 21

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service