Skip to Content
Merck
All Photos(1)

Key Documents

463515

Sigma-Aldrich

Silicon 2,3,9,10,16,17,23,24-octakis(octyloxy)-29H,31H-phthalocyanine dihydroxide

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H146N8O10Si
CAS Number:
Molecular Weight:
1600.32
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

composition

Dye content, 95%

mp

>300 °C (lit.)

λmax

682 nm

SMILES string

CCCCCCCCOc1cc2c3nc(nc4n5c(nc6nc(nc7n(c(n3)c8cc(OCCCCCCCC)c(OCCCCCCCC)cc78)[Si]5(O)O)c9cc(OCCCCCCCC)c(OCCCCCCCC)cc69)c%10cc(OCCCCCCCC)c(OCCCCCCCC)cc4%10)c2cc1OCCCCCCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service