Skip to Content
Merck
All Photos(1)

Key Documents

45930

Supelco

3,4-Dinitrobenzoic acid

analytical standard

Synonym(s):

3,4-DNBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4N2O6
CAS Number:
Molecular Weight:
212.12
Beilstein:
2055333
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

OC(=O)c1ccc(c(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H4N2O6/c10-7(11)4-1-2-5(8(12)13)6(3-4)9(14)15/h1-3H,(H,10,11)

InChI key

OMVRRHJJQILIJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dinitrobenzoic acid belongs to the class of highly polar nitroaromatic compounds, which are categorized under the group of explosives and related compounds (ERCs). ERCs are of research interest because they are identified as the potential intermediates of environmental degradation pathway of explosives that can adversely affect the ecosystem and human health.

Application

3,4-Dinitrobenzoic acid may be used as an analytical standard for the determination of the analyte in explosive contaminated waters by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of nitrophenols, nitrobenzoic acids and polar explosives by HPLC-diode array detection in ground water samples of former ammunition plants
Godejohann M, et al.
Chromatographia, 43(11-12), 612-618 (1996)
Analysis of nitrophenols and other polar nitroaromatic compounds in ammunition wastewater by high-field proton nuclear magnetic resonance (1H-NMR) spectroscopy and chromatographic methods
Preiss A, et al.
Fresenius Journal of Analytical Chemistry, 357(6), 676-683 (1997)
A Preiss et al.
Journal of chromatography. A, 1216(25), 4968-4975 (2009-05-19)
An advanced HPLC-photodiode array detection method for the determination of 12 selected highly polar nitroaromatic compounds in ground water samples of ammunition waste sites has been developed and validated. After solid-phase extraction the limits of detection were in the range
Yuko Fukuyama et al.
Journal of the American Society for Mass Spectrometry, 29(11), 2227-2236 (2018-08-01)
In in-source decay (ISD) in matrix-assisted laser desorption/ionization (MALDI)-mass spectrometry (MS), 1,5-diaminonaphthalene (1,5-DAN) is a most frequently used matrix probably due to the highly sensitive detection of fragment ions. 1,5-DAN is a reducing matrix generating c- and z-series ions by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service