Skip to Content
Merck
All Photos(1)

Key Documents

36754

Sigma-Aldrich

9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F

~99% (TLC), 10 mg/mL in methyl acetate

Synonym(s):

U-46619, U46619

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49
Beilstein:
4267334
MDL number:
UNSPSC Code:
12352200

Assay

~99% (TLC)

concentration

10 mg/mL in methyl acetate

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\[C@H]1C2CC(CO2)[C@@H]1C\C=C/CCCC(O)=O

InChI

1S/C21H34O4/c1-2-3-6-9-17(22)12-13-19-18(16-14-20(19)25-15-16)10-7-4-5-8-11-21(23)24/h4,7,12-13,16-20,22H,2-3,5-6,8-11,14-15H2,1H3,(H,23,24)/b7-4-,13-12+/t16?,17-,18-,19+,20?/m0/s1

InChI key

LQANGKSBLPMBTJ-REGKDVDGSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F

Flash Point(C)

-10 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael E Grassie et al.
The Journal of biological chemistry, 287(43), 36356-36369 (2012-09-06)
Ca(2+) sensitization of smooth muscle contraction depends upon the activities of protein kinases, including Rho-associated kinase, that phosphorylate the myosin phosphatase targeting subunit (MYPT1) at Thr(697) and/or Thr(855) (rat sequence numbering) to inhibit phosphatase activity and increase contractile force. Both
André Erdling et al.
Neuropeptides, 47(2), 85-92 (2013-02-05)
Vasoactive intestinal peptide (VIP) and pituitary adenylate cyclase activating peptide (PACAP)-containing nerves surround cerebral blood vessels. The peptides have potent vasodilator properties via smooth muscle cell receptors and activation of adenylate cyclase. The purpose of this study was to describe
Andrew W Johnson et al.
Hypertension (Dallas, Tex. : 1979), 61(2), 437-442 (2012-12-26)
Angiotensin II (Ang II) is known to promote vascular disease and hypertension in part by formation of cytokines, such as interleukin-6. However, the role of signal transducer and activator of transcription 3 (STAT3) in these processes and Ang II/interleukin-6 signaling
Bin Liu et al.
American journal of physiology. Renal physiology, 305(9), F1315-F1322 (2013-08-30)
This study was to determine whether prostacyclin [prostaglandin I2 (PGI2)] evokes mouse renal vasoconstriction and, if so, the underlying mechanism(s) and how its synthesis via cyclooxygenase-1 (COX-1) influences local vasomotor reaction. Experiments were performed on vessels from C57BL/6 mice and/or
Nitin Puri et al.
Antioxidants & redox signaling, 18(5), 471-480 (2012-08-08)
Hydrogen peroxide (H(2)O(2)), a nonradical oxidant, is employed to ascertain the role of redox mechanisms in regulation of vascular tone. Where both dilation and constriction have been reported, we examined the hypothesis that the ability of H(2)O(2) to effect vasoconstriction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service