Skip to Content
Merck
All Photos(1)

Key Documents

34762BC

Millipore

Assurance® GDS Listeria Positive Control

For use with Assurance® GDS Listeria Tq

Synonym(s):

Positive Control for PCR

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€128.00

€128.00


Estimated to ship onMay 24, 2025


Request more information

Select a Size

Change View
5 ML
€128.00

About This Item

UNSPSC Code:
41116135
NACRES:
NA.85

€128.00


Estimated to ship onMay 24, 2025


Request more information

Product Name

GDS Positive Control for Listeria, BioControl, For use with GDS Listeria

Quality Level

form

liquid

shelf life

12 mo.

packaging

vial of 5 mL (sufficient for 166 tests)

manufacturer/tradename

BioControl Systems 34762-250

storage condition

(Tightly closed)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

2-8°C

suitability

Listeria spp.

General description

GDS Positive Control for Listeria is used with GDS Listeria Tq assays, which are automated nucleic acid amplification systems for the detection of Listeria in food and environmental samples.

Application

GDS Positive Control for Listeria is used for confirmation of Listeria spp. detection in Assurance® GDS assays.

Components

One vial, 5 mL, sufficient for 166 tests

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service