Skip to Content
Merck
All Photos(1)

Documents

34524

Supelco

Ethyl butylacetylaminopropionate

PESTANAL®, analytical standard

Synonym(s):

3-[Acetyl(butyl)amino]propionic acid ethyl ester, N-Acetyl-N-butyl-β-alanine ethyl ester, Ethyl 3-(N-acetyl-N-butylamino)propionate, Ethyl 3-(N-butylacetamido)propionate, Ethyl 3-[acetyl(butyl)amino]propanoate, Ethyl N-acetyl-N-butyl-β-alaninate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO3
CAS Number:
Molecular Weight:
215.29
Beilstein:
11343172
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

InChI

1S/C11H21NO3/c1-4-6-8-12(10(3)13)9-7-11(14)15-5-2/h4-9H2,1-3H3

InChI key

VZRKEAFHFMSHCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service