Skip to Content
Merck
All Photos(1)

Key Documents

27475

Supelco

(+)-β-Citronellene

analytical standard

Synonym(s):

(S)-(+)-3,7-Dimethyl-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
1719874
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D +10±1°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437

bp

154-155 °C (lit.)

density

0.760 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](CC\C=C(\C)C)C=C

InChI

1S/C10H18/c1-5-10(4)8-6-7-9(2)3/h5,7,10H,1,6,8H2,2-4H3/t10-/m1/s1

InChI key

FUDNBFMOXDUIIE-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Citronellene is an oxygenated monoterpene and is the major constituent of essential oils isolated from different plant species. Monoterpenes are known to have bactericidal, fungicidal, antiparasitic and insecticidal properties. β-Citronellene is also used in the production of various fragrance ingredients including dihydromyrcenol and cyclodemol.
(+)-β-Citronellene is an acyclic monoterpene compound.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insecticidal effects of monoterpenes on Sitophilus zeamais Motschulsky (Coleoptera: Curculionidae).
Yildirim E, et al.
Journal of Applied Botany and Food Quality, 86, 198-204 (2013)
Synthesis of fragrance compounds from acyclic monoterpenes: Rhodium catalyzed hydroformylation and tandem hydroformylation/acetalization of linalool and $\beta$-citronellene.
Vieira CG, et al.
Applied Catalysis A: General, 466, 208-215 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service