Skip to Content
Merck
All Photos(1)

Documents

16132

Sigma-Aldrich

8-Bromoadenosine

≥99.0% (HPLC)

Synonym(s):

6-Amino-8-bromopurine riboside, 8-Bromoadenine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12BrN5O4
CAS Number:
Molecular Weight:
346.14
Beilstein:
627737
EC Number:
MDL number:
UNSPSC Code:
41106305

Assay

≥99.0% (HPLC)

optical activity

[α]20/D −33±2°, c = 1% in 1 M HCl

mp

210-212 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Nc1ncnc2n([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c(Br)nc12

InChI

1S/C10H12BrN5O4/c11-10-15-4-7(12)13-2-14-8(4)16(10)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,13,14)/t3-,5-,6-,9-/m1/s1

InChI key

VJUPMOPLUQHMLE-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Inhibits the binding of cyclic AMP and adenosine to the activated cyclic AMP/adenosine binding protein

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An adenosine 3':5'-monophosphate/adenosine binding protein from mouse liver. A study of its interaction with synthetic and naturally occurring adenosine derivatives.
P M Ueland
European journal of biochemistry, 86(1), 27-34 (1978-05-01)
N L Nolan-Sorden et al.
Analytical biochemistry, 184(2), 298-304 (1990-02-01)
2',5'-oligoadenylates known as 2-5A [px(A2'p)nA; chi = 2 or 3, n greater than or equal to 2] are produced in interferon-treated cells in response to double-stranded RNA. 2-5A binds with high affinity to a 2-5A-dependent RNase resulting in the cleavage
P Provencher et al.
Journal of steroid biochemistry, 36(6), 589-596 (1990-08-28)
Steroidogenesis was studied in guinea-pig glomerulosa-fasciculata cells maintained in primary culture for up to 7 days. The basal secretion which remained stable for the first 2 days in culture rapidly rose to reach a plateau on day 4 at levels
L H Clarson et al.
Placenta, 18(4), 333-339 (1997-05-01)
The expression of GLUT1 and GLUT3 mRNA and protein in human placental trophoblast-derived cells was investigated. A dividing choriocarcinoma derived cell line (JAr) was compared to differentiating cytotrophoblast cells, isolated from human term placenta, following 18 (mononucleate) and 66 h
Fengling Cui et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 74(4), 964-971 (2009-09-25)
This study was designed to examine the interaction of 8-bromoadenosine with human serum albumin (HSA) by fluorescence spectroscopy in combination with molecular modeling under simulative physiological conditions. The results of fluorescence measurements indicate that 8-bromoadenosine has a strong ability to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service