Skip to Content
Merck
All Photos(1)

Key Documents

12070

Sigma-Aldrich

Benzamide

purum, ≥98.0% (HPLC)

Synonym(s):

Benzoic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONH2
CAS Number:
Molecular Weight:
121.14
Beilstein:
385876
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

mp

125-128 °C (lit.)
125-128 °C

solubility

methanol: soluble 1 g/10 mL, clear, colorless to faintly yellow

SMILES string

NC(=O)c1ccccc1

InChI

1S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H2,8,9)

InChI key

KXDAEFPNCMNJSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzamide was used to study the effect of inhibitors of endonuclease and poly(ADP ribose) polymerase on anti-Fas monoclonal antibody treated Jurkat cells.

Biochem/physiol Actions

Benzamide is a specific inhibitor of poly(ADP-ribose)polymerase. It prevents glutamate- and methamphetamine-induced neurotoxicity in vitro in C57B1/6N mouse.
Inhibits poly(ADP-ribose) polymerase (PARP).

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Kun et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(23), 7219-7223 (1983-12-01)
Human fibroblasts were subjected to nutritionally induced G1 block, followed by release and subsequent entry into S phase, and exposed to nontoxic concentrations of carcinogens in early S phase. Cell transformation occurred as determined by early morphologic cell alterations, anchorage-independent
I Yamadori et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 46(1), 85-90 (1997-12-24)
We compared two methods to stain apoptotic cells, one using terminal deoxynucleotidyl transferase (TDT), the other DNA polymerase I, using leukemia cell lines treated with anti-Fas monoclonal antibody (MAb). Both TDT and polymerase I strongly reacted with fragmented nuclei of
C Cosi et al.
Brain research, 735(2), 343-348 (1996-10-07)
Previous studies have indicated that the activation of poly(ADP-ribose) polymerase (PARP), an enzyme involved in DNA plasticity-related phenomena, is an early event occurring in glutamate-induced neurotoxicity in vitro, and that inhibitors of PARP, including benzamide, are protective against both glutamate-
James M Wright et al.
The Cochrane database of systematic reviews, 4, CD001841-CD001841 (2018-04-19)
This is the first update of a review published in 2009. Sustained moderate to severe elevations in resting blood pressure leads to a critically important clinical question: What class of drug to use first-line? This review attempted to answer that
M H Gschwend et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 831(1-2), 132-139 (2006-01-03)
The development of a validated liquid chromatography-tandem mass spectrometry (LC-MS/MS) method with positive electrospray ionisation (ESI(+)) and multiple reaction monitoring (MRM) for the selective and sensitive bioanalytical determination of amisulpride, a substituted benzamide derivative, in human plasma is described. Plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service