Skip to Content
Merck
All Photos(1)

Key Documents

11514

Supelco

Hydroxylamine hydrochloride

≥99.9995% (metals basis)

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein:
3539763
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

Assay

≥99.9995% (metals basis)

form

solid

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.005 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤1 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.1 mg/kg
Cs: ≤0.01 mg/kg
Cu: ≤0.2 mg/kg
Fe: ≤0.5 mg/kg
Ga: ≤0.01 mg/kg
Hg: ≤0.001 mg/kg
In: ≤0.01 mg/kg
Ir: ≤0.01 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.2 mg/kg
Mn: ≤0.05 mg/kg
Mo: ≤0.01 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.05 mg/kg
Os: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pd: ≤0.01 mg/kg
Pt: ≤0.1 mg/kg
Rb: ≤0.01 mg/kg
Rh: ≤0.01 mg/kg
Ru: ≤0.01 mg/kg
Sb: ≤0.01 mg/kg
Se: ≤0.01 mg/kg
Sn: ≤0.05 mg/kg
Sr: ≤0.01 mg/kg
Tl: ≤0.01 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.2 mg/kg

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bereket Y Oquare et al.
Bioconjugate chemistry, 19(11), 2196-2204 (2008-10-04)
We report the design and synthesis of an orthogonally protected peptide nucleic acid (PNA) building block, Fmoc-PNA-U'-(Dde)-OH, and its use in the construction of PNA FRET probes. This building block allows for the post-synthetic attachment of reporter groups to the
Daniel F Dwyer et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 529-539 (2014-06-15)
Papain, a cysteine protease allergen with inherent adjuvant activity, induces potent IL-4 expression by T cells in the popliteal lymph nodes of mice following footpad immunization. In this study, we identify a novel, non-BCR-mediated capacity for B cells to rapidly
Anke Bill et al.
PloS one, 9(6), e97973-e97973 (2014-06-03)
The human prostacyclin receptor (hIP receptor) is a seven-transmembrane G protein-coupled receptor (GPCR) that plays a critical role in vascular smooth muscle relaxation and platelet aggregation. hIP receptor dysfunction has been implicated in numerous cardiovascular abnormalities, including myocardial infarction, hypertension
Yuan Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 10(22), 4770-4777 (2014-08-20)
Chiral plasmonic assemblies with strong and tunable chiroptical activity are emerging materials yet challenging to fabricate. Moreover, shell-programmed chiroptical regulation is really rare. Here, the chiroptical activity of core-shell (CS) nanoparticles (NPs) heterodimers (HDs) with different types and thicknesses of
Simeon K Adesina et al.
Drug development and industrial pharmacy, 40(11), 1547-1556 (2013-09-26)
Nanoparticle size is important in drug delivery. Clearance of nanoparticles by cells of the reticuloendothelial system has been reported to increase with increase in particle size. Further, nanoparticles should be small enough to avoid lung or spleen filtering effects. Endocytosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service