Skip to Content
Merck
All Photos(1)

Key Documents

10815

Sigma-Aldrich

Ethyl β-apo-8′-carotenoate (trans)

≥80% (TLC)

Synonym(s):

Ethyl 8′-apo-β,ψ-caroten-8′-oate, all-trans-Carophyll yellow

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H44O2
CAS Number:
Molecular Weight:
460.69
Beilstein:
2021845
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥80% (TLC)

impurities

≤0.001% heavy metals

ign. residue

≤0.1%

loss

≤0.1% loss on drying

mp

134-136 °C

solubility

chloroform: 10 mg/mL at 25 °C, clear, intense red-orange

storage temp.

−20°C

SMILES string

CCOC(=O)\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

Other Notes

Internal standard in: Individual carotenoid determinations in human plasma by HPLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Franke et al.
Journal of chromatography, 614(1), 43-57 (1993-04-21)
A method is described for the synthesis of new carotenoids by transesterification of ethyl-beta-apo-8'-carotenoate. The rate of transesterification was temperature-dependent with highest yields using primary alcohols. Reaction conditions were found avoiding Z isomerization. The structures of the new products were
C.C. Tangney
Journal of Liquid Chromatography, 7, 2611-2611 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service