Skip to Content
Merck
All Photos(1)

Key Documents

04059

Sigma-Aldrich

Fmoc-Inp-OH

≥97.0% (HPLC)

Synonym(s):

1-Fmoc-piperidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
Beilstein:
7722550
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

mp

~190 °C (dec.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)C1CCN(CC1)C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C21H21NO4/c23-20(24)14-9-11-22(12-10-14)21(25)26-13-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-8,14,19H,9-13H2,(H,23,24)

InChI key

OJYOOHSQOIDDOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for anti-thrombotic peptide libraries

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.P. Combs et al.
Journal of the American Chemical Society, 118, 287-287 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service