Skip to Content
Merck
All Photos(1)

Documents

04003

Supelco

Tetradecane

certified reference material, TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12CH3
CAS Number:
Molecular Weight:
198.39
Beilstein:
1733859
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

6.83 (vs air)

vapor pressure

1 mmHg ( 76.4 °C)

product line

TraceCERT®

autoignition temp.

455 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.429 (lit.)

bp

252-254 °C (lit.)

mp

5.5 °C (lit.)

density

0.762 g/mL at 20 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCC

InChI

1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3

InChI key

BGHCVCJVXZWKCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service