Skip to Content
Merck
All Photos(1)

Key Documents

24-5650

Sigma-Aldrich

Potassium tartrate dibasic hemihydrate

JIS special grade, ≥99.0%

Synonym(s):

L-(+)-Tartaric acid dipotassium salt, Dipotassium tartrate hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOOC(CHOH)2COOK · 0.5H2O
CAS Number:
Molecular Weight:
235.28
Beilstein:
6121656
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.0%
grade:
JIS special grade
form:
crystalline
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

7.0-8.5 (25 °C, 235.3 g/L)

SMILES string

O.[K+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2K.H2O/c5-1(3(7)8)2(6)4(9)10;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;1H2/q;2*+1;/p-2/t1-,2-;;;/m1.../s1

InChI key

BUCMCXRUIRJPKS-QXMMYYGRSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service