Skip to Content
Merck
All Photos(1)

Documents

13-1730

Sigma-Aldrich

Hydrochloric acid

0.1 M

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:

product name

Hydrochloric acid solution, 0.1 M

vapor pressure

190 mmHg ( 25 °C)

form

liquid

availability

available only in Japan

concentration

0.1 M
1/10 N

bp

100 °C (lit.)

density

1 g/cm3 at 20 °C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc: This study explores a novel approach using hydrochloric acid solution in the synthesis process aimed at valorizing waste materials. The article demonstrates a chemical strategy that enhances resource recovery, significantly contributing to waste minimization in industrial practices (Morgovan et al., 2024).

  • Contactless conductivity sensor employing moist paper as absorbent for in-situ detection of generated carbon dioxide gas: The study integrates hydrochloric acid solutions in the development of a novel, cost-effective contactless conductivity sensor. This sensor uses a simple paper-based system for the in-situ monitoring of gases, providing a significant tool in environmental monitoring and analysis (Nacapricha et al., 2020).


Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal
Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Davide R Ceratti et al.
Advanced materials (Deerfield Beach, Fla.), 27(34), 4958-4962 (2015-07-23)
A new, simple bi-phasic dip-coating method is developed. This method is considered as a great improvement of the technique for research, development and production, since expensive, rare, harmful, or time-evolving solutions can now be easily deposited on large surfaces and
Rebecca J Ford et al.
The Biochemical journal, 468(1), 125-132 (2015-03-06)
Metformin is the mainstay therapy for type 2 diabetes (T2D) and many patients also take salicylate-based drugs [i.e., aspirin (ASA)] for cardioprotection. Metformin and salicylate both increase AMP-activated protein kinase (AMPK) activity but by distinct mechanisms, with metformin altering cellular
Jay G Hosking et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 1005-1015 (2014-10-21)
Successful decision making often requires weighing a given option's costs against its associated benefits, an ability that appears perturbed in virtually every severe mental illness. Animal models of such cost/benefit decision making overwhelmingly implicate mesolimbic dopamine in our willingness to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service