Skip to Content
Merck
All Photos(1)

Key Documents

05-6190

Sigma-Aldrich

Copper standard solution

suitable for atomic absorption spectrometry, 1000 ppm Cu, 1 mg/mL Cu

Synonym(s):

Copper(II) sulfate solution, Cupric sulfate standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

concentration

1 mg/mL Cu
1000 ppm Cu

suitability

suitable for atomic absorption spectrometry

storage temp.

15-25°C

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonella Amore et al.
FEMS microbiology letters, 337(2), 155-163 (2012-10-12)
In silico analyses of several laccase promoter sequences have shown the presence of many different responsive elements differentially distributed along the promoter sequences. Analysis of Pleurotus ostreatus laccase promoter poxa1b extending around 1400-bp upstream of the start codon showed the
Eun A Choi et al.
International journal of molecular medicine, 31(1), 154-162 (2012-11-07)
Egg white-chalcanthite (EWCC) is a mixture of egg white and chalcanthite prepared by roasting chalcanthite (which is a natural mineral mainly composed of CuSO4•5H2O) to the point of dehydration, pulverizing the dehydrated chalcanthite and then mixing the pulverized chalcanthite to
M Seder-Colomina et al.
Ecotoxicology (London, England), 22(1), 199-205 (2012-11-10)
Microbial mats are coastal ecosystems that consist mainly of cyanobacteria, primary producers in these habitats that play an important role in stabilising delta sediments. However, these ecosystems are subject to various kinds of pollution, including metal contamination, placing their survival
Xu-Chen Cao et al.
PloS one, 8(2), e56735-e56735 (2013-03-08)
The aquaporin (AQP) family consists of a number of small integral membrane proteins that transport water and glycerol. AQPs are critical for trans-epithelial fluid transport. Recent reports demonstrated that AQPs, particularly AQP1 and AQP5, are expressed in high grade tumor
Ang Li et al.
Oncology reports, 29(2), 535-540 (2012-11-21)
Aquaporins (AQPs) are a family of small, integral membrane proteins that have been shown to play an important role in tumor development and metastasis. Several studies have demonstrated that expression of AQP3 contributes to the enhanced migration of epithelial cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service