Skip to Content
Merck
All Photos(1)

Key Documents

01-4340

Sigma-Aldrich

Ammonium hydroxide solution

JIS special grade, 25.0-27.9% NH3

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

form

liquid

expl. lim.

27 %

availability

available only in Japan

concentration

25.0-27.9% NH3

pH

11.7 (20 °C)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Cequier et al.
Analytica chimica acta, 845, 98-104 (2014-09-10)
Organophosphate triesters are common flame retardants used in a wide variety of consumer products from which they can migrate and pollute the indoor environment. Humans may thus be continuously exposed to several organophosphate triesters which might be a risk for
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Dominic J Hare et al.
ACS chemical neuroscience, 6(3), 398-402 (2015-01-15)
Plasma iron levels are decreased in Alzheimer's disease (AD) and associated with an idiopathic anemia. We examined iron-binding plasma proteins from AD patients and healthy controls from the Australian Imaging, Biomarkers and Lifestyle (AIBL) Flagship Study of Ageing using size
Marilia Freitas Calmon et al.
Colloids and surfaces. B, Biointerfaces, 100, 177-184 (2012-07-07)
Magnetic iron oxide nanoparticles (magnetite) (MNPs) were prepared using different organic and inorganic bases. Strong inorganic base (KOH) and organic bases (NH(4)OH and 1,4-diazabicyclo[2.2.2]octane (DABCO)) were used in the syntheses of the MNPs. The MNPs were characterized by X-ray diffraction
Zhuo Li et al.
Biosensors & bioelectronics, 70, 246-253 (2015-04-02)
A magnetic and fluorescent nano-composite was prepared. It comprised of a core of Fe3O4 nanoparticles (NPs), a silica shell and satellitic Au nano-clusters (AuNCs) capped with bovine serum albumin (BSA). This nano-composite has many desirable properties, e.g. magnetism, red emission

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service