Skip to Content
Merck
All Photos(2)

Key Documents

V159-15GS

Fluid Thioglycollate Medium (FTM)

vial of 15 mL, pkg of 4 units, suitable for sterility testing, sterilization compatible with autoclavable

Synonym(s):

FTM, Thioglycollate Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
Pricing and availability is not currently available.

material

glass vial (20ml vial)

Quality Level

Agency

EP 2.6.13
JP 4.06
USP 71

Sterilization Compatibility

autoclavable compatible

form

liquid

packaging

vial of 15 mL
pkg of 4 units

technique(s)

microbiological culture: suitable
sterility testing: suitable

pH

6.9±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid Thioglycollate Medium (FTM) is a complex medium designed for the cultivation and isolation of fastidious aerobic and anaerobic microorganisms present in sterility test samples of pharmaceutical industry.
The anaerobic atmosphere is created by reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment. The presence of beef extract adds to the carbon source and other essential vitamins in the medium to support the growth of microorganisms present in the sample.
The presence of agar may reduce the rapid diffusion of oxygen and lead to slight turbidity in larger volumes (filled bottles). Resazurin indicator in the medium indicates the reduction potential. An increase in oxygen concentration is indicated through a color-change reaction to pink.

Application

Fluid Thioglycollate Medium (FTM) is mainly used as a sterility testing medium for sterile pharmaceutical samples. to cultivate and isolate anaerobic and microaerophilic microbes.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service