SYMBDSL03
Steritest® Symbio Drain Tray Support for 3 canisters, ISL Pump
Streamlines the workflow, Increases Safety
Synonym(s):
Canister holder
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
316L stainless steel (Drain Tray Support)
EPDM rubber seal
Agency
complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)
manufacturer/tradename
Steritest®
application(s)
pharmaceutical
sterility testing
compatibility
for use with Steritest® Symbio Pump
shipped in
ambient
General description
Steritest® Symbio drain tray support includes the stainless steel canister holder and quick connector. This tray is designed for 3 canisters and the usage of ISL pump.
Application
Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service