SCYT-91K-PXBK14
Ovine Cytokine/Chemokine Panel 1 14 Plex
Immuno-Oncology Multiplex Assay
Synonym(s):
14 Plex Chemokine Panel, Ovine Cytokine Assay, Ovine Cytokine Panel
About This Item
species reactivity
mouse
packaging
pkg of 1 ea
manufacturer/tradename
Milliplex®
assay range
accuracy: 101
(Galectin-1)
accuracy: 102
(PD-L2)
accuracy: 103
(CD80/B7-1)
accuracy: 103
(IFNγ)
accuracy: 104
(CTLA-4/CD152)
accuracy: 104
(TLR-2)
accuracy: 105
(4-1BBL/TNFSF9)
accuracy: 105
(BCA-1/CXCL-13)
accuracy: 105
(CD25/IL-2Rα)
accuracy: 105
(IL-10)
accuracy: 105
(LAG-3)
accuracy: 106
(5’-NT/CD73)
accuracy: 106
(PD-1)
accuracy: 106
(TIM-3)
accuracy: 106
(TNFα)
accuracy: 107
(CD137/4-1BB)
accuracy: 107
(CD27)
accuracy: 108
( Galectin-3)
accuracy: 108
(E-Cadherin)
accuracy: 108
(HVEM)
accuracy: 109
(CD40)
accuracy: 109
(GITR)
accuracy: 110
(B7-H2/ICOSL)
accuracy: 110
(CD226/DNAM-1)
accuracy: 111
(BTLA)
accuracy: 94
(GranzymeB)
accuracy: 95
(B7-H3/CD276)
accuracy: 96
(PD-L1)
sensitivity: 14 - 10000 pg/mL
(4-1BBL/TNFSF9)
standard curve range: 1.4 - 1000 pg/mL
(HVEM)
standard curve range: 1.4 - 1000 pg/mL
(IFNγ)
standard curve range: 11 - 8000 pg/mL
(IL-10)
standard curve range: 137 - 100000 pg/mL
(5’-NT/CD73)
standard curve range: 14 - 10000 pg/mL
(CD226/DNAM-1)
standard curve range: 14 - 10000 pg/mL
(CD40)
standard curve range: 14 - 10000 pg/mL
(PD-L1)
standard curve range: 2.7 - 2000 pg/mL
(CD25/IL-2Rα)
standard curve range: 21 - 15000 pg/mL
(BTLA)
standard curve range: 27 - 20000 pg/mL
(B7-H2/ICOSL)
standard curve range: 27 - 20000 pg/mL
(BCA-1/CXCL-13)
standard curve range: 3.4 - 2500 ng/mL
(Galectin-1)
standard curve range: 3.4 - 2500 ng/mL
(B7-H3/CD276)
standard curve range: 34.3 - 25000 pg/mL
(CD80/B7-1)
standard curve range: 342 - 250000 pg/mL
(E-Cadherin)
standard curve range: 4.1 - 3000 pg/mL
(TNFα)
standard curve range: 41 - 30000 pg/mL
(GITR)
standard curve range: 41 - 30000 pg/mL
(GranzymeB)
standard curve range: 41 - 30000 pg/mL
(Galectin-3)
standard curve range: 412 - 300000 pg/mL
(LAG-3)
standard curve range: 412 - 300000 pg/mL
(TLR-2)
standard curve range: 5.5 - 4000 pg/mL
(CD137/4-1BB )
standard curve range: 5.5 - 4000 pg/mL
(CTLA-4/CD152)
standard curve range: 5.5 - 4000 pg/mL
(TIM-3)
standard curve range: 55 - 40000 pg/mL
(CD27)
standard curve range: 55 - 40000 pg/mL
(PD-1)
standard curve range: 6.9 - 5000 pg/mL
(PD-L2)
inter-assay cv: <20%
intra-assay cv: <10%
technique(s)
multiplexing: suitable
compatibility
configured for Premixed
detection method
fluorometric (Luminex® xMAP®)
shipped in
wet ice
storage temp.
2-8°C
Related Categories
General description
MILLIPLEX® Mouse Immuno-Oncology Checkpoint Protein Panel 1 is a 28-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, cell culture supernatant, cell lysate, and tissue/tumor extract samples: 4-1BBL/TNFSF9, 5′-NT/CD73, B7-H2/ICOSL, B7-H3/CD276, BCA-1/CXCL-13, BTLA, CD137/4-1BB, CD226/DNAM-1, CD25/IL-2Rα, CD27, CD40, CD80/B7-1, CTLA-4/CD152, E-cadherin, Galectin-1, Galectin-3, GITR, Granzyme B, HVEM, IFNγ, IL-10, LAG-3, PD-1, PD-L1, PD-L2, TIM-3, TLR-2, and TNFα.
Panel Type: MAGNETIC Circulating Cancer
Specificity
Application
This is an overnight incubation assay (16-18 hours at 2-8ºC).
A maximum of 25 μL per well 1:2 diluted serum or plasma can be used.
Tissue culture samples may be run neat or appropriately diluted in control medium.
Note: Best results using cell/tissue lysate samples may require diluting in assay buffer while testing similarly diluted lysis buffer for matrix effects.
Research Category: Cancer/Immuno-oncology
Analytes:
4-1BBL/TNFSF9
5’-NT/CD73
B7-H2/ICOSL
B7-H3/CD276
BCA-1/CXCL-13
BTLA
CD137/4-1BB
CD226/DNAM-1
CD25/IL-2Rα
CD27
CD40
CD80/B7-1
CTLA-4/CD152
E-cadherin
Galectin-1
Galectin-3
GITR
Granzyme B
HVEM
IFNγ
IL-10
LAG-3
PD-1
PD-L1
PD-L2
TIM-3
TLR-2
TNFα
Features and Benefits
Packaging
Other Notes
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service