Skip to Content
Merck
All Photos(2)

Key Documents

PHWP14250

Millipore

MCE Membrane Filter, 0.3 μm Pore Size

MF-Millipore, filter diam. 142 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 0.3 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

142 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 50 discs

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

3 L/min-cm2 air flow rate
32 mL/min-cm2 water flow rate
75 °C max. temp.

diam.

142 mm

filter diam.

142 mm

thickness

150 μm

gravimetric extractables

2%

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.3 μm pore size
77 % porosity

bubble point

≥2.4 bar, air with water at 23 °C

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: PHWP

Application

  • Bioassays
  • Air monitoring
  • Particle monitoring
  • Particle removal

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service