Skip to Content
Merck
All Photos(1)

Documents

PHIX174

Millipore

VIRAPREP Coliphage(ΦX174) Positive Control

new

ISO 10705-2, suitable for water monitoring, positive for microbiological tests (Somatic coliphages)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116132
NACRES:
NA.74

Agency

ISO 10705-2

shelf life

12 mo.

packaging

pack of 1 × 20 tubes

suitability

positive for microbiological tests (Somatic coliphages)

application(s)

water monitoring

storage temp.

−20°C

General description

VIRAPREP Coliphage(ΦX174) Positive Control is designed as a dehydrated positive quality control standard for coliform enumeration.
The ΦX174 Positive Control is a freeze-dried water soluble biological material, containing a specified number of ΦX174 phages.

Application

VIRAPREP Coliphage(ΦX174) Positive Control is used with VIRAPREP kit for the enumeration of somatic coliphages in drinking water samples.

Features and Benefits

The positive control can be used without defrosting.
It can be rehydrated using sterile water.
It contains around 100 PFU (Plaque Forming Units) per vial

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service