Skip to Content
Merck
All Photos(2)

Documents

P3C010C10

Millipore

Pellicon® 3 Cassette with Ultracel® Membrane

filtration area 1.14 m2, NMWCO 10 kDa

Synonym(s):

Pellicon® 3 Cassette with Ultracel 10 kDa Membrane, C screen, 1.14 m2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602

material

TPE seal
composite regenerated cellulose (CRC)
polypropylene screen
stainless steel cap (for end; polypropylene-encapsulated)

Quality Level

feature

holdup volume 170 mL (in feed channel)

manufacturer/tradename

Pellicon®

parameter

30 psi max. reverse press. exposure at 25 °C (for 24 hours)
4-6 L/min-m2 recirculation rate
40 psi max. transmembrane pressure at 4-50 °C
6.9 bar max. inlet pressure (100 psi)
80 psi max. transmembrane pressure at 4-40 °C

technique(s)

analytical sample preparation: suitable
buffer exchange: suitable
protein purification: suitable (concentration)

H

3.8 cm (1.51 in.)

L

20.6 cm (8.1 in.)

W

17.8 cm (7 in.)

filtration area

1.14 m2

membrane area

1.14 m2

gravimetric extractables

≤1500 mg/m2

matrix

Ultracel® (Membrane)

pore size

10 kDa NMWCO

pH

2-13

Looking for similar products? Visit Product Comparison Guide

General description

Pellicon® 3 filtration devices are advanced, high-performance cassettes ideal for today’s higher titer therapeutic antibodies as well as the more demanding filtration processes that require higher operating pressures, temperatures and caustic cleaning regimes.

The Pellicon® 3 Cassette with Ultracel® Membrane is designed for the filtration of therapeutic proteins (such as monoclonal antibodies) and recombinant and non-recombinant proteins.

This P3C010C10 Pellicon® 3 ultrafiltration device features a pore size of 10 kDa NMWCO, a filtration area of 1.14 m2, a PLCGC membrane, and dimensions measuring 2.4 cm (H) x 17.8 cm (W) x 20.6 cm (L).

Features and Benefits

  • Rugged & reliable – ideally suited for today′s more demanding filtration processes
  • High performance – produces excellent resolution, yields and back-pressure resistance
  • Optimum product recovery – thanks to proven membrane technology
  • Void-free technology – leading-edge membranes resolve virtually any separation challenge
  • Defect-free, superior adhesion – Ultracel®uses composite membranes cast on a microporous substrate
  • Automated manufacturing – delivers unbeatable performance, consistency and reliability
  • Easy installation & care – simple to install and maintain

Other Notes

These products are manufactured in a facility which adheres to Good Manufacturing Practices.

Analysis Note

Gravimetric Extractables: after 24 hours in water at controlled room temperature

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service