material
EPDM rubber
EPDM rubber device (ethylene propylene diene monomer)
Quality Level
feature
hydrophobic (Surface)
parameter
-30-140 °C temp. range (Short term use)
-6-(-1) bar pressure range
weight
0.1 kg (0.220 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany