Skip to Content
Merck
All Photos(1)

Key Documents

MXQLAB001

Millipore

Milliflex® Quantum Funnel Labels

For use with Milliflex® Quantum system

Synonym(s):

traceability labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.35

manufacturer/tradename

Milliflex®

application(s)

pharmaceutical

compatibility

for use with Milliflex® Quantum Systems

shipped in

ambient

General description

Funnel Labels are designed to be used with the Milliflex® Quantum system. Milliflex® Quantum is a fluorescence-based technology for the quantitative and rapid detection of microorganisms in filterable samples. The universal enzymatic fluorescent staining is non-destructive, allowing the identification of the contaminant following a positive result. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. Funnel Labels with relevant QC data are used to ensure traceability during a Milliflex® filtration test.

Application

The Milliflex® Quantum Funnel Labels are used to ensure traceability during the Milliflex® filtration test of filtrable samples, including in-process and final product samples (pharmaceuticals and water monitoring).

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service