MMA125
Millicell® µ-Angiogenesis Inhibition Assay Kit
About This Item
Recommended Products
species reactivity
rat, vertebrates, human
technique(s)
activity assay: suitable
cell culture | mammalian: suitable
detection method
fluorometric
Mode of action
| inhibits
General description
Tubular formation is a multi-step process involving cell adhesion, migration, differentiation, and growth. Millipore′s TM=["Millicell"] µ-Angiogenesis Inhibition Assay provides an efficient system for the rapid and accurate identification of factors that inhibit tube formation of endothelial cells. The µ-Angiogenesis slide, with enhanced optical capabilities in a microscale format, allows for real time and continuous monitoring of tube formation in a simplifed and reproducible manner.
Components
Storage and Stability
Legal Information
Disclaimer
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service