Skip to Content
Merck
All Photos(1)

Documents

MABT1492

Sigma-Aldrich

Anti-Desmoplakin Antibody, clone 20B6

clone 20B6, from mouse

Synonym(s):

DP, 250/210 kDa paraneoplastic pemphigus antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20B6, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DSP(1832)

General description

Desmoplakin (UniProt: P15924; also known as DP, 250/210 kDa paraneoplastic pemphigus antigen) is encoded by the DSP gene (Gene ID:1832) in human. Desmoplakin is a major high molecular weight homodimeric protein that is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes, which are prominent adhesive junctions present between many epithelial cells and cardiomyocytes. Desmoplakin is the most abundant of the desmosomal proteins and plays a key role in tissue morphogenesis and maintaining the integrity of tissue. Desmoplakin is composed of an N-terminal plakin domain, a central alpha-helical coiled-coil rod domain, and a C-terminal intermediate filament-binding domain. The N-terminal domain is required for desmosome association and the C-terminal binds to desmin. Three isoforms for Desmoplakin have been reported that are produced by alternative splicing. Desmoplakin 2 and 3 lack most of the central rod domain. Desmoplakin 1 is the dominant isoform in the cardiac tissue. Desmoplakin is phosphorylated by protein kinase A on serine 2849 and this phosphorylation affects its association with epidermal, simple cytokeratins, and VIM intermediate filaments. Mutations in DSP gene have been linked to DCWHK (Cardiomyopathy, dilated, with woolly hair and keratoderma) that is characterized by a generalized striate keratoderma particularly affecting the palmoplantar epidermis, woolly hair, and dilated left ventricular cardiomyopathy. Mice deficient in Desmoplakin perish around the time of implantation (E6.5) and show fewer desmosomes.

Specificity

Clone 20B6 detects human Desmoplankin. It targets an epitope with in the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 863 amino acids from the C-terminal half of human Desmoplakin.

Application

Anti-Desmoplakin, clone 20B6, Cat. No. MABT1492, is a mouse monoclonal antibody that detects human Desmoplakin and has been tested for use in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A representative lot detected Desmoplakin in Western Blotting applications (Sobolik-Delmaire, T., et. al. (2006). J Biol Chem. 281(25):16962-70; Hall, C., et. al. (2009). Cell Commun Adhes. 16(1-3):15-27; Roberts, B.J., et. al. (2011). Exp Cell Res. 317(20):2814-22).

Immunocytochemistry Analysis: A representative lot detected Desmoplakin in Immunocytochemistry applications (Roberts, B.J., et. al. (2011). Exp Cell Res. 317(20):2814-22; Sobolik-Delmaire, T., et. al. (2006). J Biol Chem. 281(25):16962-70; Roberts, B.J., et. al. (2016). J Biol Chem. 291(48):24857-24865).

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected Desmoplakin in A431 cells.

Target description

331.77 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwabena A Badu-Nkansah et al.
Molecular biology of the cell, 31(11), 1140-1153 (2020-04-03)
Desmosomes are cell-cell adhesions necessary for the maintenance of tissue integrity in the skin and heart. While the core components of desmosomes have been identified, peripheral components that modulate canonical or noncanonical desmosome functions still remain largely unexplored. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service