Skip to Content
Merck
All Photos(2)

Documents

MABS1882

Sigma-Aldrich

Anti-TAOK3 Antibody, clone 1H9.1

clone 1H9.1, from mouse

Synonym(s):

Serine/threonine-protein kinase TAO3, EC:2.7.11.1, Cutaneous T-cell lymphoma-associated antigen HD-CL-09, CTCL-associated antigen HD-CL-09, Dendritic cell-derived protein kinase, JNK/SAPK-inhibitory kinase, Jun kinase-inhibitory kinase, JIK, Kinase from

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H9.1, monoclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TAOK3(51347)

General description

Serine/threonine-protein kinase TAO3 (UniProt: Q9H2K8; also known as EC:2.7.11.1, Cutaneous T-cell lymphoma-associated antigen HD-CL-09, CTCL-associated antigen HD-CL-09, Dendritic cell-derived protein kinase, JNK/SAPK-inhibitory kinase, Jun kinase-inhibitory kinase, JIK, Kinase from chicken homolog A, hKFC-A, Thousand and one amino acid protein 3) is encoded by the TAOK3 (also known as DPK, JIK, KDS, MAP3K18) gene (Gene ID: 51347) in human. TAO kinase 3 is a member of the STE20 subfamily of the STE Ser/Thr protein kinases. It is a serine/threonine-protein kinase that acts as a regulator of the p38/MAPK14 stress-activated MAPK cascade and of the MAPK8/JNK cascade. In response to DNA damage at the G2/M phase, it is phosphorylated at Ser324 by ATM and acts as an activator of the p38/MAPK14 stress-activated MAPK cascade. It is shown to inhibit basal activity of MAPK8/JNK cascade and also diminishes its activation in response epidermal growth factor. TAO kinase 3 is highly expressed in peripheral blood leukocytes, thymus, spleen, kidney, skeletal muscle, heart, and liver.

Specificity

Clone 1H9.1 specifically detects Serine/threonine-protein kinase TAO3 (TAOK3) in K562 cells. It targets an epitope of 16 amino acids in the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminus end of human Serine/threonine-protein kinase TAO3 (TAOK3).

Application

Detect TAOK3 using this mouse monoclonal Anti-TAOK3 Antibody, clone 1H9.1, Cat. No. MABS1882. It has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TAOK3 in human testis, human small intestine, and human pancreas.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected TAOK3 in 10 µg of Raw264.7 cell lysate.
Research Category
Signaling

Quality

Evaluated by Western Blotting in K562 cells.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TAOK3 in 10 µg of K562 cell lysate.

Target description

~120 kDa observed, 105.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service