Skip to Content
Merck
All Photos(1)

Key Documents

MABN14

Sigma-Aldrich

Anti-phospho TDP-43 (Ser409/Ser410) Antibody, clone 1D3

clone 1D3, from rat

Synonym(s):

TAR DNA-binding protein 43, TDP-43, TDP43, TAR DNA binding protein-43, Phosphorylated TDP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D3, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer409/pSer410)

Gene Information

human ... TARDBP(23435)

General description

TAR DNA-binding protein 43 (TDP-43) belongs to the hnRNP protein family and plays an important role in transcription, pre-mRNA splicing, mRNA stability and mRNA transport. It is involved in splicing of the apolipoprotein A-II and cystic fibrosis transmembrane gene. This protein is highly expressed in the pancreas, placenta, lung, genital tract and spleen. Mutations in TDP-43 have been associated with amyotrophic lateral sclerosis, frontotemporal dementia, Parkinson′s disease and Alzheimer′s disease.

Specificity

This antibody recognizes TDP-43 when phosphorylated at Ser409/Ser410.

Immunogen

Epitope: Ser409/Ser410
Ovalbumin-conjugated linear peptide corresponding to human TDP-43 phosphorylated at Ser409/Ser410.

Application

Anti-phospho TDP-43 (Ser409/Ser410) Antibody, clone 1D3 is an antibody against phospho TDP-43 (Ser409/Ser410) for use in WB, ELISA, IH.
Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in hippocampus from FTLD-U brain tissue. (Neumann, M., et al. (2009). Acta Neuropathol. 117:137–149.)

ELISA Analysis: A previous lot was used by an independent laboratory in ELISA. (Neumann, M., et al. (2009). Acta Neuropathol. 117:137–149.)
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blot in human placenta cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected phosphorylated TDP-43 on 10 µg of human placenta cell lysate.

Target description

50/25 kDa Observed
1) The 25-kDa C-terminal fragment is more prone to phosphorylation at Ser409/Ser410 than full-length TDP-43. (Zhang, Y.J., et al. (2009). PNAS. 106:7607-7612.)
2) 50-52 kDa observed MW . (Buratti, E., et al. (2001). The EMBO Journal 20:1774-1784.)

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placenta cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick J Bosque et al.
PloS one, 8(5), e62301-e62301 (2013-05-25)
The transactive response DNA-binding protein (TDP-43) is a major component of the abnormal intracellular inclusions that occur in two common neurodegenerative diseases of humans: (1) a subtype of frontotemporal lobar degeneration and (2) amyotrophic lateral sclerosis. Genetics, experiments in cultured
Zhi Huang et al.
Nature communications, 14(1), 2747-2747 (2023-05-13)
Resilience to Alzheimer's disease is an uncommon combination of high disease burden without dementia that offers valuable insights into limiting clinical impact. Here we assessed 43 research participants meeting stringent criteria, 11 healthy controls, 12 resilience to Alzheimer's disease and
Pol Andrés-Benito et al.
Journal of neuropathology and experimental neurology, 78(5), 416-425 (2019-04-03)
Dyneins are major components of microtubules. Dynein assembly is modulated by a heterogeneous group of dynein axonemal assembly factors (DNAAFs). The present study analyzes dynein axonemal assembly factor 1 (DNAAF1) and leucine-rich repeat-containing protein 50 (LRRC50), the corresponding encoded protein
Jacob R Mann et al.
Neuron, 102(2), 321-338 (2019-03-04)
TDP-43 proteinopathy is a pathological hallmark of amyotrophic lateral sclerosis and frontotemporal dementia where cytoplasmic TDP-43 inclusions are observed within degenerating regions of patient postmortem tissue. The mechanism by which TDP-43 aggregates has remained elusive due to technological limitations, which
Akila Weerasekera et al.
NeuroImage. Clinical, 27, 102327-102327 (2020-07-13)
Currently TAR DNA binding protein 43 (TDP-43) pathology, underlying Amyotrophic Lateral Sclerosis (ALS), remains poorly understood which hinders both clinical diagnosis and drug discovery efforts. To better comprehend the disease pathophysiology, positron emission tomography (PET) and multi-parametric magnetic resonance imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service