MABF3161
Anti-β-Catenin Antibody, clone 12F7
Synonym(s):
b-Cat
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
12F7, monoclonal
mol wt
calculated mol wt 85.44 kDa
observed mol wt ~95 kDa
purified by
using protein G
species reactivity
human
species reactivity (predicted by homology)
zebrafish, bovine, Xenopus, chicken, rat, mouse
packaging
antibody small pack of 100 μg
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
chicken ... B-Cat(395964)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in A431 cell lysates.
Western Blotting Analysis: A 1:125 dilution of this antibody detected β-Catenin in A431 cell lysates.
Tested Applications
Immunohistochemistry Applications: A representative lot detected β-Catenin in Immunohistochemistry applications (Qu, Y., et al. (2016). Proc. Natl. Acad. Sci. USA.113(33); 9339-44).
Western Blotting Analysis: A representative lot detected β-Catenin in Western Blotting applications (Johnson, K.R., et al. (1993). Exp. Cell Res. 207(2); 252-60).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service