Skip to Content
Merck
All Photos(1)

Key Documents

MABF145B

Sigma-Aldrich

Anti-TCR beta chain (mouse), APC, clone H57-597 Antibody

clone H57-597, from hamster(Armenian), Allophycocyanin conjugate

Synonym(s):

Pre T-cell antigen receptor alpha, TCR beta chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster (Armenian)

conjugate

Allophycocyanin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

H57-597, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Tcrb(21577)

General description

The H57-597 antibody is specific for the beta chain of the mouse T cell Receptor (TCR). This cell surface protein combines with a second protein chain (alpha chain) to form the alpha-beta TCR that is expressed by NK1.1+ thymocytes, NKT cells, and the majority of peripheral T cells. A small number of T cells may express an alternative heteromer of gamma and delta protein chains, known as the gamma-delta TCR. These receptors participate in a complex with CD3, and with the co-receptors CD4 or CD8, to recognize and respond to antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. The H57-597 antibody is used as a phenotypic marker for T cells expressing the alpha-beta TCR. It is also widely used to cross-link surface TCR and thereby mimic TCR-mediated cell activation or induction of apoptosis. The antibody does not cross-react with cells expressing the gamma-delta TCR.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-TCR beta chain (mouse), APC, clone H57-597 Antibody is an antibody against TCR beta chain for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected TCR beta chain in one million C57Bl/6 splenocytes.

Physical form

Purified Armenian Hamster monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service