Skip to Content
Merck
All Photos(1)

Key Documents

MAB944

Sigma-Aldrich

Anti-Coxsackievirus B1 Antibody, clone 334-4A-8F-3F

clone 334-4A-8F-3F, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

334-4A-8F-3F, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with Coxsackie virus B1. Neutralizes Coxsackie virus B1 Conn 5 strain at a titer of less than 1:16.

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Suggested starting dilution for IFA is 1:1000.

Dilute with buffer pH 7.5 -8.0 containing protein or other stabilizing medium. Final working dilutions must be determined by end user.
This Anti-Coxsackievirus B1 Antibody, clone 334-4A-8F-3F is validated for use in IF for the detection of Coxsackievirus B1.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remi Villenave et al.
PloS one, 12(2), e0169412-e0169412 (2017-02-02)
Analysis of enterovirus infection is difficult in animals because they express different virus receptors than humans, and static cell culture systems do not reproduce the physical complexity of the human intestinal epithelium. Here, using coxsackievirus B1 (CVB1) as a prototype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service