Skip to Content
Merck
All Photos(1)

Documents

MAB80143

Sigma-Aldrich

Anti-Norwalk Virus Antibody, clone 2A2-6E10-1D8

clone 2A2-6E10-1D8, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A2-6E10-1D8, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

General description

Norwalk virus is a major cause of outbreaks of gastroenteritis, affecting primarily older children and adults.

Specificity

Specific to a recombinant Norwalk Virus capsid protein

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Norwalk Virus Antibody, clone 2A2-6E10-1D8 is validated for use in ELISA, WB for the detection of Norwalk Virus.

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norwalk virus infection of volunteers: new insights based on improved assays
Graham, D Y, et al
The Journal of Infectious Diseases, 170, 34-43 (1994)
K Y Green et al.
Journal of clinical microbiology, 31(8), 2185-2191 (1993-08-01)
Since the discovery of the Norwalk virus (NV) by immune electron microscopy (IEM) in 1972, serologic studies with this virus have relied on particle-positive fecal material from infected volunteers as the source of antigen because it has not been possible
Michael J Springer et al.
Vaccine, 34(12), 1452-1458 (2016-02-14)
Norovirus is the primary cause of viral gastroenteritis in humans with multiple genotypes currently circulating worldwide. The development of a successful norovirus vaccine is contingent on its ability to induce both systemic and mucosal antibody responses against a wide range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service