Skip to Content
Merck
All Photos(1)

Key Documents

MAB416

Sigma-Aldrich

Anti-Interferon-β Antibody, clone A1

clone A1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€356.00

€356.00


Estimated to ship onMay 27, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1982

Request a Bulk Order

Select a Size

Change View
100 μG
€356.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€356.00


Estimated to ship onMay 27, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1982

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgGκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IFNB1(3456)

Specificity

Human interferon-beta (IFN-beta). Recognizes amino acids 30-47 of IFN-beta. No cross-reactivity with Human IFN-alpha or IFN-gamma.

Immunogen

Human interferon beta.

Application

Anti-Interferon-β Antibody, clone A1 detects level of Interferon-β & has been published & validated for use in ELISA & WB.
Immunoblotting

ELISA

Neutralization of IFN-beta bioactivity; 90% reduction of bioactivity at 1.4 μg/mL for recombinant IFN-beta and 2.7 μg/mL for natural IFN-beta. Assay method used is provided in Grossberg, S.E. et al, Manual of Clinical Immunology.

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Format: Purified
Hybridoma culture grown in low IgG FBS (3%), Protein G purified. Liquid in 50 mM Na Acetate, pH 6.5, 50 mM NaCl, 4% sucrose, 0.1% Tween20, no preservatives added

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to twelve months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service